Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piribedil dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10mg
50mg
Description
Specifications
Quantity | 50mg |
Formula Weight | 371.27 |
Percent Purity | >99% |
Chemical Name or Material | Piribedil dihydrochloride |
Safety and Handling
Recommended Storage : Store at Room Temperature