Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pirlindole mesylate, Tocris Bioscience™

Catalog Number 072450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
207572-66-5
C16H22N2O3S
322.423
PVUYTBQAVAYLDE-BTQNPOSSSA-N
2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
124080994
CC1=CC2=C(C=C1)N3CCNC4C3=C2CCC4.CS(=O)(=O)O
This item is not returnable. View return policy
207572-66-5
C16H22N2O3S
322.423
PVUYTBQAVAYLDE-BTQNPOSSSA-N
2,3,3a,4,5,6-Hexahydro-8-methyl-1H-pyrazino[3,2,1-j,k]carbazole mesylate
124080994
CC1=CC2=C(C=C1)N3CCNC4C3=C2CCC4.CS(=O)(=O)O

MAO-A inhibitor

A highly selective reversible inhibitor of monoamine oxidase type A.
Quantity 50 mg
Formula Weight 322.42
Percent Purity >98%
Chemical Name or Material Pirlindole mesylate
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit