Learn More
PISD Mouse anti-Human, Clone: OTI6C3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807333
Catalog No. 50-166-8224
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
Specifications
Specifications
PISD
Monoclonal
Unconjugated
PISD
Lyophilized
PISD
Human recombit protein fragment corresponding to amino acids 80-340 of human PISD produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
OTI6C3
PBS with 8% trehalose and no preservative; pH 7.3
Q9UG56
DJ858B16, PSD, PSDC, PSSC, dJ858B16.2
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23761