Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Piston For Titration Sensor Interchangable Burettes
GSA_VA

Replacement part for DV1005 or DV1020 titration burettes.

Supplier:  Mettler Toledo™ 51107117

 View more versions of this product

Catalog No. 01-911-113


Only null left
Add to Cart

Description

Description

For 20mL titration sensor interchangable burettes.

Compliance

ISO 8655-3

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Piston
20 mL Burettes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.