Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIT 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective PIP3 antagonist. Blocks the binding of PIP3 to the pleckstrin homology (PH) domain of Akt (IC50 = 31.03 μM). Inhibits cancer cell survival and induces apoptosis by inhibition of PIP3-dependent PI 3-kinase/Akt signaling. Exhibits antitumor activity in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 351.76 |
Percent Purity | >99% |
Chemical Name or Material | PIT 1 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction