Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110201
Description
Immunogen sequence: MDSPEIRELE KFANEFKVRR IKLGYTQTNV GEALAAVHGS EFSQTTICRF ENLQLSFKNA CKLKAILSKW LEEAEQVGAL YNEKVGANER KRKRRTTISI AAKDALERHF GEQNKPSSQE IMRMAEELNL EKEVVRVWFC NRRQREKRVK.
PIT1 is a pituitary-specific transcription factor responsible for pituitary development and hormone expression in mammals and is a member of the POU family of transcription factors that regulate mammalian development. The POU family is so named because the first 3 members identified were PIT1 and OCT1 of mammals, and Unc-86 of C. elegans. PIT1 contains 2 protein domains, termed POU-specific and POU-homeo, which are both necessary for high affinity DNA binding on genes encoding growth hormone and prolactin. PIT1 is also important for regulation of the genes encoding prolactin and thyroid-stimulating hormone beta subunit by thyrotropin-releasing hormone and cyclic AMP.
Specifications
PIT1 | |
Polyclonal | |
Unconjugated | |
POU1F1 | |
CPHD1; dw; dwarf; GHF1; GHF-1; GHF1A; growth hormone factor 1; Hmp1; homeodomain protein for Growth Hormone (GH); PIT1; pit-1; Pit1-rs1; PIT1Z; Pituary specific transcription factor 1 (growth hormone factor 1); pituitary specific transcription factor 1; pituitary specific transcription factor 1, related sequence 1; pituitary-specific positive transcription factor 1; pituitary-specific positive transcription factor 1 variant w; POU class 1 homeobox 1; POU domain, class 1, transcription factor 1; POU domain, class 1, transcription factor 1 (Pit1, growth hormone factor 1); POU1F1; POU1F1a; transcription factor Pit-1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5449 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.87 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P28069 | |
POU1F1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 124-273 of human POU1F1 (NP_0002971?). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction