Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITPN Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549316
Description
Predicted to react with bovine and rabbit based on sequence homology.
Catalyzes the transfer of PtdIns and phosphatidylcholine between membranes.Specifications
PITPN | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q00169, P53810, P16446 | |
PITPNA | |
KLH conjugated synthetic peptide between 243-270 amino acids from the C-terminal region of human PITPNA. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PITPNA | |
Phosphatidylinositol transfer protein alpha isoform, PI-TP-alpha, PtdIns transfer protein alpha, PtdInsTP alpha, PITPNA, PITPN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18738, 29525, 5306 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction