Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPNM1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548828

Catalog No. PA548828

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Regulates RHOA activity, and plays a role in cytoskeleton remodeling. Necessary for normal completion of cytokinesis. Plays a role in maintaining normal diacylglycerol levels in the Golgi apparatus. Binds phosphatidyl inositol phosphates (in vitro). May catalyze the transfer of phosphatidylinositol and phosphatidylcholine between membranes. Necessary for maintaining the normal structure of the endoplasmic reticulum and the Golgi apparatus. Required for protein export from the endoplasmic reticulum and the Golgi. Binds calcium ions.
Specifications

Specifications

PITPNM1
Polyclonal
PBS with 0.09% sodium azide
O00562, O35954
Membrane-associated phosphatidylinositol transfer protein 1, Drosophila retinal degeneration B homolog, Phosphatidylinositol transfer protein, membrane-associated 1, PITPnm 1, Pyk2 N-terminal domain-interacting receptor 2, NIR-2, PITPNM1, DRES9, NIR2, PITPNM
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18739, 9600
Western Blot
Unconjugated
PITPNM1
Liquid
PITPNM1
KLH conjugated synthetic peptide between 143-171 amino acids from the N-terminal region of human PITPNM1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit