Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPNM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548616

Catalog No. PA548616


Add to cart

Description

Description

Catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes (in vitro). Binds calcium ions.
Specifications

Specifications

PITPNM2
Polyclonal
PBS with 0.09% sodium azide
Q9BZ72
Membrane-associated phosphatidylinositol transfer protein 2, Phosphatidylinositol transfer protein, membrane-associated 2, PITPnm 2, Pyk2 N-terminal domain-interacting receptor 3, NIR-3, PITPNM2, KIAA1457, NIR3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57605
Western Blot
Unconjugated
PITPNM2
Liquid
PITPNM2
KLH conjugated synthetic peptide between 639-668 amino acids from the Central region of human PITPNM2.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit