Learn More
Aviva Systems Biology Corp PITX3 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP33032_P050
This is a rabbit polyclonal antibody against PITX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PITX3. Alternate names: CTPP4, MGC12766, PTX3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54857