Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pivaloyl hydrazide, 98+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
42826-42-6
C5H12N2O
116.16
MFCD01310839
OARJXUPBZNUYBG-UHFFFAOYSA-N
2,2-dimethylpropionic acid hydrazide,pivalic acid hydrazide,pivalohydrazide,2,2-dimethylpropanohydrazide,pivaloylhydrazine,2,2-dimethyl-propionic acid hydrazide,propanoic acid, 2,2-dimethyl-, hydrazide,pivalic acid, hydrazide,2,2-dimethylpropionicacidhydrazide,trimethylacethydrazide
545147
2,2-dimethylpropanehydrazide
CC(C)(C)C(=O)NN
42826-42-6
C5H12N2O
116.16
MFCD01310839
OARJXUPBZNUYBG-UHFFFAOYSA-N
2,2-dimethylpropionic acid hydrazide,pivalic acid hydrazide,pivalohydrazide,2,2-dimethylpropanohydrazide,pivaloylhydrazine,2,2-dimethyl-propionic acid hydrazide,propanoic acid, 2,2-dimethyl-, hydrazide,pivalic acid, hydrazide,2,2-dimethylpropionicacidhydrazide,trimethylacethydrazide
545147
2,2-dimethylpropanehydrazide
CC(C)(C)C(=O)NN
Boiling Point 95°C to 110°C (1.0 mbar)
Melting Point 65°C to 67°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 1g
Assay Percent Range 98% min. (GC)
Linear Formula (CH3)3CC(O)NHNH2
Solubility Information Solubility in water: .. Other solubilities: soluble in alcohols
Formula Weight 116.16
Physical Form Crystals
Percent Purity 98+%
Chemical Name or Material Pivaloyl hydrazide
Show More Show Less

GHS H Statement:
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Risk of explosion if heated under confinement.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning