Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 156591AP150UL
Description
Hiwi is a member of the Piwi/Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34-hematopoietic cells, suggesting a role for this gene in the maintece of hematopoietic stem cells.Specifications
PIWIL1 | |
Polyclonal | |
Unconjugated | |
PIWIL1 | |
HIWI, MIWI, PIWI, piwi like 1 (Drosophila), Piwi like protein 1, PIWIL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57749, 9271 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96J94, Q9JMB7 | |
Piwil1 | |
PIWIL1 Fusion Protein Ag8056 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction