Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL4 Polyclonal Antibody
PIWIL4 Polyclonal Antibody
Supplier: Thermo Scientific PA557731
Description
PIWIL4 Polyclonal Antibody for IHC

Specifications
PIWIL4 | |
Polyclonal | |
Unconjugated | |
PIWIL4 | |
9230101H05Rik; HIWI2; mAgo5; Miwi2; PIWI; piwi like RNA-mediated gene silencing 4; Piwil4; piwi-like 4; piwi-like homolog 4; piwi-like protein 4; piwi-like RNA-mediated gene silencing 4; testis tissue sperm-binding protein Li 85P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
143689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3Z4 | |
PIWIL4 | |
Recombinant protein corresponding to Human PIWIL4. Recombinant protein control fragment (Product #RP-97318). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction