Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIWIL4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549710

Catalog No. PA549710


Add to cart

Description

Description

The antibody detects endogenous level of total PIWIL4 protein.

PIWIL4 belongs to the Argonaute family of proteins, which function in development and maintece of germline stem cells (Sasaki et al., 2003 [PubMed 12906857]).
Specifications

Specifications

PIWIL4
Polyclonal
Unconjugated
PIWIL4
Liquid
PIWIL4
Fusion protein corresponding to a region derived from 546-838 amino acids of human piwi-like RNA-mediated gene silencing 4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
1.1 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q7Z3Z4
HIWI2, PIWI
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
143689
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit