Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PK 44 phosphate, Tocris Bioscience™

Catalog Number 414510
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
1017682-66-4
C17H19F5N7O5P
527.349
IRIGBESJZBGJOS-DDWIOCJRSA-N
pk 44 phosphate, 3r-3-amino-4-6,7-difluoro-1h-indazol-3-yl-1-5,6-dihydro-3-trifluoromethyl-1,2,4-triazolo 4,3-a pyrazin-7 8h-yl-1-butanone phosphate
90488917
(3R)-3-amino-4-(6,7-difluoro-2H-indazol-3-yl)-1-[3-(trifluoromethyl)-6,8-dihydro-5H-[1,2,4]triazolo[4,3-a]pyrazin-7-yl]butan-1-one;phosphoric acid
C1CN2C(=NN=C2C(F)(F)F)CN1C(=O)CC(CC3=C4C=CC(=C(C4=NN3)F)F)N.OP(=O)(O)O
This item is not returnable. View return policy
1017682-66-4
C17H19F5N7O5P
527.349
IRIGBESJZBGJOS-DDWIOCJRSA-N
pk 44 phosphate, 3r-3-amino-4-6,7-difluoro-1h-indazol-3-yl-1-5,6-dihydro-3-trifluoromethyl-1,2,4-triazolo 4,3-a pyrazin-7 8h-yl-1-butanone phosphate
90488917
(3R)-3-amino-4-(6,7-difluoro-2H-indazol-3-yl)-1-[3-(trifluoromethyl)-6,8-dihydro-5H-[1,2,4]triazolo[4,3-a]pyrazin-7-yl]butan-1-one;phosphoric acid
C1CN2C(=NN=C2C(F)(F)F)CN1C(=O)CC(CC3=C4C=CC(=C(C4=NN3)F)F)N.OP(=O)(O)O

Potent and selective dipeptidyl peptidase IV (DPP-IV) inhibitor

Potent inhibitor of dipeptidyl peptidase IV (DPP-IV) (IC50 = 15.8 nM). Displays >1000-fold selectivity for DPP-IV over DPP-8 and DPP-9. Improves glucose tolerance in a mouse oral glucose tolerance assay.
Formula Weight 527.34
Percent Purity >98%
Quantity 10 mg
Chemical Name or Material PK 44 phosphate
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit