Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA C alpha Mouse anti-Human, Biotin, Clone: 7H3A4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237246B

Catalog No. NBP237246B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PKA C alpha
7H3A4
Western Blot, ELISA
Purified
PRKACA
Purified recombinant fragment of human PRKACA (AA: 1-120) expressed in E. Coli.
Protein A or G purified
Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary
5566
Western Blot, ELISA
Biotin
PBS with 0.05% Sodium Azide
cAMP-dependent protein kinase catalytic subunit alpha, EC 2.7.11, EC 2.7.11.11, MGC102831, MGC48865, PKA C-alpha, PKACA, protein kinase A catalytic subunit, protein kinase, cAMP-dependent, catalytic, alpha
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit