Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA R2 Mouse anti-Human, Mouse, Rat, FITC, Clone: OTI1F8, Novus Biologicals™

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP273428F

Catalog No. NBP273428F


Add to cart

Description

Description

PKA R2 Monoclonal specifically detects PKA R2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PKA R2
Monoclonal
FITC
PBS with 0.05% Sodium Azide
PRKAR2A
Full length recombinant protein of human PKA R2 (NP_004148) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI1F8
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit
Mouse
Affinity Purified
Phospho Specific, Signal Transduction
5576.0
Store at 4°C in the dark.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit