Learn More
PKA R2 Mouse anti-Human, Porcine, Bovine, Alexa Fluor™ 350, Clone: RS30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP229843AF350
Catalog No. NP229843A30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PKA R2 Monoclonal antibody specifically detects PKA R2 in Human,Porcine,Bovine samples. It is validated for Western Blot,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
PKA R2
RS30
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Regulatory subunit of cAMP-dependant protein kinase type II, alpha and beta.
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
PRKAR2A
Purified regulatory subunit of cAMP-dependant protein kinase type II from bovine brain.
0.1 mL
Primary
5576
Bovine, Human, Porcine