Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKA R2 Mouse anti-Human, Porcine, Bovine, Alexa Fluor™ 750, Clone: RS30, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP229843AF750

Catalog No. NP229843A70

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PKA R2 Monoclonal antibody specifically detects PKA R2 in Human, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

PKA R2
RS30
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
PRKAR2A
Purified regulatory subunit of cAMP-dependant protein kinase type II from bovine brain.
Protein A or G purified
Phospho Specific, Signal Transduction
Primary
5576.0
Bovine, Human, Porcine
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
protein kinase, cAMP-dependent, regulatory, type II, alpha, RII-alpha subunit
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Regulatory subunit of cAMP-dependant protein kinase type II, alpha and beta.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit