Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKA RI beta Mouse anti-Human, Mouse, Rat, Biotin, Clone: OTI2A3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273430B
Description
PKA RI beta Monoclonal specifically detects PKA RI beta in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PKA RI beta | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI2A3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
PRKAR1B | |
Full length recombinant protein of human PKA regulatory subunit I beta (NP_002726) produced in HEK293T cells. | |
0.1 mL | |
5575.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title