Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC 412, Tocris Bioscience™

Catalog Number 29921
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
120685-11-2
C35H30N4O4
570.65
MFCD12828879
IIQIEMHSDLLZQA-QZPVEUDVSA-N
[9S-(9α,10β,11β,13α)]-N-(2,3,10,11,12,13-Hexahydro-10-methoxy-9-methyl-1-oxo-9,13-epoxy-1H,9H-diindolo[1,2,3-gh:3',2',1'-lm]pyrro
101631538
N-[(15R,16S,17R,18S)-17-methoxy-17,18-dimethyl-5-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaen-16-yl]-N-methylbenzamide
CO[C@]1(C)[C@@H]([C@H]2O[C@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O)N(C)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
120685-11-2
C35H30N4O4
570.65
MFCD12828879
IIQIEMHSDLLZQA-QZPVEUDVSA-N
[9S-(9α,10β,11β,13α)]-N-(2,3,10,11,12,13-Hexahydro-10-methoxy-9-methyl-1-oxo-9,13-epoxy-1H,9H-diindolo[1,2,3-gh:3',2',1'-lm]pyrro
101631538
N-[(15R,16S,17R,18S)-17-methoxy-17,18-dimethyl-5-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8,10,12,20(25),21,23-nonaen-16-yl]-N-methylbenzamide
CO[C@]1(C)[C@@H]([C@H]2O[C@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O)N(C)C(=O)C1=CC=CC=C1

Protein kinase C inhibitor

Broad spectrum protein kinase inhibitor. Inhibits conventional PKC isoforms (α, α, γ), PDFRα, VEGFR2, Syk, PKCη, Flk-1, Flt3, Cdk1/B, PKA, c-Kit, c-Fgr, c-Src, VEGFR1, EGFR and MARK (microtubule affinity regulating kinase). Displays potent antitumor activity. Identified as targeting human host cell proteins that interact with SARS-CoV-2.
Quantity 1 mg
Formula Weight 570.64
Percent Purity >98%
Chemical Name or Material PKC 412
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit