Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC alpha Mouse anti-Human, Mouse, Rat, Clone: 133, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen PKC alpha
Clone 133
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Western Blot, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122142
View Documents
Novus Biologicals
NBP30733420UG
20 μg
Each for $138.37
Add to cart
 
NB122141
View Documents
Novus Biologicals
NBP30733410UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

PKC alpha Monoclonal antibody specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications

Specifications

PKC alpha
0.2 mg/ml
Western Blot, Immunohistochemistry (Paraffin)
Purified
RUO
Human, Mouse, Rat
AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha
Full-length protein corresponding to PKC alpha. Recognizes sequence PQFVHPILQSAV at the C terminus at PKC alpha
Monoclonal
Protein A or G purified
133
Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cancer, Cellular Markers, mTOR Pathway, Phospho Specific, Signal Transduction, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
5578
IgG2a Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit