Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC alpha (T638)/PKC beta II (T641) Rabbit anti-Human, Mouse, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

Manufacturer:  Abnova Corporation PAB30171

Catalog No. 89-033-729


Add to cart

Description

Description

Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human PKC alpha/PKC beta II.

  • High specificity
  • High reactivity
Specifications

Specifications

PKC alpha (T638)/PKC beta II (T641)
Unconjugated
ELISA (0.05-0.2 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
P17252; P05771
AAG6|MGC129900|MGC129901|PKC-alpha|PKCA|PRKACA
Rabbit
IgG
40 ug
Store at 4°C is stable for 2-3 weeks. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of PKC alpha only when phosphorylated at threonine 638 and PKC beta II only when phosphorylated at threonine 641.
ELISA, Western Blot
Rabbit polyclonal antibody raised against partial synthetic phosphopeptide of human PKC alpha/PKC beta II.
In 0.5 mg/mL PBS, pH 7.4 (30% glycerol, 0.02% sodium azide).
Liquid
PRKCA
A synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T638 of human PKC alpha and T641 of human PKC beta II.
Affinity purification
RUO
Primary
5578|5579
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)