Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKC beta 2 Mouse anti-Human, Mouse, Clone: 469827, R&D Systems™

Mouse Monoclonal Antibody

$116.82 - $320.50

Specifications

Antigen PKC beta 2
Clone 469827
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB43781100 View Documents R&D Systems
MAB43781100
100 ug Each for $320.50
Add to cart
 
MAB43781SP View Documents R&D Systems
MAB43781SP
25 ug Each for $116.82
Add to cart
 
Description

Description

PKC beta 2 Monoclonal antibody specifically detects PKC beta 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

PKC beta 2
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL
Unconjugated
Mouse
Human, Mouse
P05771
E. coli-derived recombinant human PKC beta 2 Lys607-Ser673 Accession # P05771
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
469827
Immunohistochemistry, Western Blot
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
5579
IgG1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human PKC beta 2 in direct ELISAs. Detects human and mouse PKC beta 2 in Western blots.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit