Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKC epsilon Mouse anti-Human, Mouse, Rat, Monkey, Primate, HRP, Clone: OTI4G8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273438H
Description
PKC epsilon Monoclonal specifically detects PKC epsilon in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PKC epsilon | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PRKCE | |
Full length recombinant protein of human PKC epsilon (NP_005391) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI4G8 | |
Western Blot, Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.13, MGC125656, nPKC-epsilon, PKCEMGC125657, protein kinase C epsilon type, protein kinase C, epsilon | |
Mouse | |
Affinity Purified | |
Apoptosis, Cancer, Phospho Specific, Signal Transduction | |
5581.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title