Learn More
PKD2L2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSP00010W
Catalog No. PIOSP00010W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PKD2L2
Polyclonal
PKD2L2
PKD2L2, PK2L2, Polycystin-L2
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9NZM6
PKD2L2
A synthetic peptide from the 4th cytoplasmic loop of human Polycystic kidney disease 2-like 2 protein
RUO
27039
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations