Learn More
Azzota Corp PKI-587 DFS Item
Manufacturer:  Azzota Corp C1270
PKI-587 (Gedatolisib) is an agent targeting the phosphatidylinositol 3 kinase (PI3K) and mammalian target of rapamycin (mTOR) in the PI3K/mTOR signaling pathway, with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3171