Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKIA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA538638
Description
PKIA Polyclonal Antibody for ICC/IF, IHC (P)
The protein encoded by this gene is a member of the cAMP-dependent protein kinase (PKA) inhibitor family. This protein was demonstrated to interact with and inhibit the activities of both C alpha and C beta catalytic subunits of the PKA. Alternatively spliced transcript variants encoding the same protein have been reported.
Specifications
PKIA | |
Polyclonal | |
Unconjugated | |
PKIA | |
AI415001; cAMP-dependent protein kinase inhibitor alpha; cAMP-dependent protein kinase inhibitor, muscle/brain isoform; PKIA; PKIalpha; PKI-alpha; PRKACN1; protein kinase (cAMP-dependent, catalytic) inhibitor alpha; protein kinase inhibitor, alpha | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114906, 18767, 5569 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P61925, P63248, P63249 | |
PKIA | |
A synthetic peptide derived from the internal region of human IPKA. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction