Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PKIB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556371
Description
Immunogen sequence: MHILFVDVAM RTDSSKMTDV ESGVANFASS ARAGRRNALP DIQSSAATDG TSDLPLKLEA LSVKEDA Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 72%.
This gene encodes a member of the cAMP-dependent protein kinase inhibitor family. The encoded protein may play a role in the protein kinase A (PKA) pathway by interacting with the catalytic subunit of PKA, and overexpression of this gene may play a role in prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Jul 2012].
Specifications
PKIB | |
Polyclonal | |
Unconjugated | |
PKIB | |
cAMP-dependent protein kinase (catalytic subunit binding) inhibtor 2; cAMP-dependent protein kinase inhibitor 2; cAMP-dependent protein kinase inhibitor beta; cAMP-dependent protein kinase inhibitor bet-like protein; cAMP-dependent protein kinase inhibitor, testis isoform; im:7054531; Pkib; Pkibeta; PKI-beta; PKINH; PRKACN2; protein kinase (cAMP dependent, catalytic) inhibitor beta; protein kinase (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, cAMP dependent, catalytic; protein kinase inhibitor beta, cAMP dependent, testis specific; protein kinase inhibitor, beta; protein kinase, cAMP-dependent, catalytic, inhibitor 2; RATPKINH; si:dkey-150h13.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5570 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C010 | |
PKIB | |
Recombinant protein corresponding to Human PKIB. Recombinant protein control fragment (Product #RP-94851). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction