Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM2 Mouse anti-Human, Alexa Fluor™ 750, Clone: 1B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242686AF750

Catalog No. NP242686A70


Add to cart

Description

Description

PKM2 Monoclonal antibody specifically detects PKM2 in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

PKM2
1B10
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
PKM
Recombinant human PKM2 (1-531aa) purified from E.coli.
100 μL
Primary
5315
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit