Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM2 Rabbit anti-Human, Mouse, Rat, Bovine, PE/Atto594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP148308PEATT594

Catalog No. N148308P594

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PKM2 Polyclonal antibody specifically detects PKM2 in Human, Mouse, Rat, Bovine samples. It is validated for Flow Cytometry.
Specifications

Specifications

PKM2
PE/ATTO 594
PBS with 0.05% Sodium Azide
CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Polyclonal
Bovine, Human, Mouse, Rat
Flow Cytometry
Flow Cytometry
Affinity Purified, Conjugated
PKM
A synthetic peptide made to the internal region of human PKM2 protein (within residues 350-450). [Swiss-Prot: P14618]
Immunogen affinity purified
Cancer, Lipid and Metabolism
Primary
5315.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit