Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548703

Catalog No. PA548703

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Contributes to invasiveness in maligt prostate cancer.
Specifications

Specifications

Pkn3
Polyclonal
PBS with 0.09% sodium azide
Q6P5Z2, Q8K045
Serine/threonine-protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, Pkn3, Pknbeta
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
263803, 29941
Western Blot
Unconjugated
PKN3
Liquid
PKN3
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of mouse Pkn3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit