Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN3 Rabbit anti-Human, Mouse, DyLight 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130102V

Catalog No. NBP130102V


Add to cart

Description

Description

PKN3 Polyclonal antibody specifically detects PKN3 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

PKN3
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.11, EC 2.7.11.13, PKNbeta, protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, RP11-545E17.1, serine/threonine-protein kinase N3
Rabbit
IgG
100 ul
Protein Kinase
Primary
29941
Western Blot
Western Blot
Affinity Purified
PKN3
maps to a region between residue 225 and 275 of human protein kinase N3 using the numbering given in entry NP_037487.2
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only