Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKP3 Mouse anti-Human, Clone: 23E3/4, Abnova™

Mouse monoclonal antibody raised against human PKP3.

Manufacturer:  Abnova Corporation MAB12683

Catalog No. 89-934-774


Add to cart

Description

Description

This gene encodes a member of the arm-repeat (armadillo) and plakophilin gene families. Plakophilin proteins contain numerous armadillo repeats, localize to cell desmosomes and nuclei, and participate in linking cadherins to intermediate filaments in the cytoskeleton. This protein may act in cellular desmosome-dependent adhesion and signaling pathways. [provided by RefSeq]

Sequence: CFTPQSRRLRELPLAADALTF
Specifications

Specifications

PKP3
23E3/4
Mouse monoclonal antibody raised against human PKP3.
In PBS (0.09% sodium azide).
PKP3
A synthetic peptide corresponding to amino acids 570-589 of human PKP3.
Protein G purification
RUO
Primary
11187
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunocytochemistry Immunohistochemistry (Frozen sections) (1:25-1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Immunoprecipitation Western Blot (1:100-1:1000) The optimal working dilution should be d
Liquid
Mouse
IgG2b
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit