Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKR Mouse anti-Human, FITC, Clone: 6H3A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237242F

Catalog No. NBP237242F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PKR
6H3A10
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
EIF2AK2
Purified recombinant fragment of human EIF2AK2 (AA: 329-551) expressed in E. Coli.
Protein A or G purified
Apoptosis, Phospho Specific, Signal Transduction
Primary
5610
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
FITC
PBS with 0.05% Sodium Azide
double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit