Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKR (p Thr446) Rabbit anti-Human, Clone: SY230, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP267426

Catalog No. NBP267426


Add to Cart

Description

Description

PKR Monoclonal specifically detects PKR in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

PKR (p Thr446)
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Chromatin Immunoprecipitation, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated
Rabbit
Protein A purified
Apoptosis, Phospho Specific, Signal Transduction
5610
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
ChIP Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
SY230
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
EIF2AK2
Synthetic phospho-peptide corresponding to residues surrounding Thr446 of Human PKR aa 421-470/ 551. (SwissProt: P19525 Human)
100 μL
Primary
Human
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit