Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLA1A Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282024IR
This item is not returnable.
View return policy
Description
PLA1A Polyclonal antibody specifically detects PLA1A in Human, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
PLA1A | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 755 | |
EC 3.1.1, EC 3.1.1.-, NMD, phosphatidylserine-specific phospholipase A1alpha, phospholipase A1 member A, PS-PLA1, PSPLA1Phosphatidylserine-specific phospholipase A1 | |
PLA1A antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human PLA1A. The immunogen is located within amino acids 380 - 430 of PLA1A. Amino Acid Squence: KFKFQSSNRVWKKDRTT | |
0.1 mL | |
Immunology | |
51365 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction