Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G12A Mouse anti-Human, Clone: 7C7C9, Abnova™

Mouse monoclonal antibody raised against partial recombinant human PLA2G12A.

Manufacturer:  Abnova Corporation MAB12253

Catalog No. 89-934-776


Add to cart

Description

Description

Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s (Gelb et al., 2000 [PubMed 11031251]).[supplied by OMIM]

Specifications

Specifications

PLA2G12A
7C7C9
Mouse monoclonal antibody raised against partial recombinant human PLA2G12A.
In PBS (0.05% sodium azide)
GXII/PLA2G12/ROSSY
Mouse
IgG1
RUO
Primary
81579
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000) Flow Cytometry (1:200-1:400) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
PLA2G12A
Recombinant protein corresponding to amino acids 21-189 of human PLA2G12A.
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit