Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLA2G12A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551326
Description
The antibody detects endogenous levels of total PLA2G12A protein.
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s.
Specifications
PLA2G12A | |
Polyclonal | |
Unconjugated | |
PLA2G12A | |
2310004B05Rik; FKSG38; group XII secreted phospholipase A2; group XII-1 phospholipase A2; group XIIA secreted phospholipase A2; group XIIA secretory phospholipase A2; GXII; GXII sPLA2; GXII-1; GXII-1-PLA2; mGXII-1; phosphatidylcholine 2-acylhydrolase 12A; phospholipase A2 group XIIA; phospholipase A2, group XII; phospholipase A2, group XIIA; Pla2g12; PLA2G12A; Rossy; sPLA2-XII; UNQ2519/PRO6012 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
81579 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9BZM1 | |
PLA2G12A | |
Fusion protein of human PLA2G12A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction