Learn More
PLA2G1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158431AP
Catalog No. 50-173-0523
Add to cart
Description
Description
Phospholipase A2 catalyzes the release of fatty acids from glycero-3-phosphocholines. The best known varieties are the digestive enzymes secreted as zymogens by the pancreas of mammals. Sequences of pancreatic PLA2 enzymes from a variety of mammals have been reported. One striking feature of these enzymes is their close homology to venom phospholipases of snakes. Other forms of PLA2 have been isolated from brain, liver, lung, spleen, intestine, macrophages, leukocytes, erythrocytes, inflammatory exudates, chondrocytes, and platelets.
Specifications
Specifications
PLA2G1B
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P04054, P04055, Q9Z0Y2
Group IB phospholipase A2, Phospholipase A2, PLA2, PLA2A, PLA2G1B, PPLA2
Rabbit
IgG
150 μL
-20°C
Primary
18778, 29526, 5319
Western Blot
Unconjugated
PLA2G1B
Liquid
PLA2G1B
PLA2G1B Fusion Protein Ag8603
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat