Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G4B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen PLA2G4B
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123737
View Documents
Novus Biologicals
NBP309418100UL
100 μg
Each for $352.89
Add to cart
 
NB123738
View Documents
Novus Biologicals
NBP30941825UL
25 μg
Each for $106.19
Add to cart
 
Description

Description

PLA2G4B Polyclonal antibody specifically detects PLA2G4B in Human samples. It is validated for Western Blot
Specifications

Specifications

PLA2G4B
Western Blot
Purified
RUO
Human
FLJ20543, FLJ20656, FLJ20807, FLJ77014, FLJ78330, jmjC domain-containing protein 7, jumonji domain containing 7, Jumonji domain-containing protein 7
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PLA2G4B. Peptide sequence LQLLGRFCQEQGIPFPPISPSPEEQLQPRECHTFSDPTCPGAPAVLHFPL
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Signal Transduction
PBS buffer, 2% sucrose
8681
Polyclonal
Affinity purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit