Learn More
PLA2G7/PAF-AH/Lp-PLA2 Rabbit anti-Human, Alexa Fluor 532, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP299645AF532
Catalog No. NB062249
Add to cart
Description
Description
PLA2G7/PAF-AH/Lp-PLA2 Polyclonal specifically detects PLA2G7/PAF-AH/Lp-PLA2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
PLA2G7/PAF-AH/Lp-PLA2
Alexa Fluor 532
50mM Sodium Borate
2-acetyl-1-alkylglycerophosphocholine esterase, EC 3.1.1, EC 3.1.1.47,1-alkyl-2-acetylglycerophosphocholine esterase, Group-VIIA phospholipase A2, gVIIA-PLA2, LDL-associated phospholipase A2, LDL-PLA(2), LDL-PLA2, lipoprotein-associated phospholipase A2, LP-PLA2, PAF acetylhydrolase, PAFAHPAF 2-acylhydrolase, phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma), platelet-activating factor acetylhydrolase
Produced in rabbits immunized with purified, recombinant PLA2G7/PAF-AH/Lp-PLA2 (Q13093-1; Met1-Asn441).
Affinity purified
RUO
Primary
7941
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human