Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2R1 Rabbit anti-Human,Mouse, Unconjugated, Clone: 1D5, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen PLA2R1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB012625MI
View Documents
MilliporeSigma
ZRB01265725UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB012657MI
View Documents
MilliporeSigma
ZRB0126574X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

PLA2R1 Monoclonal antibody specifically detects PLA2R1 in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

PLA2R1
Unconjugated
Rabbit
Secretory phospholipase A2 receptor;PLA2-R;180 kDa secretory phospholipase A2 receptor;C-type lectin domain family 13-member C;M-type receptor.
GST/His-tagged recombinant fragment corresponding to 136 amino acids from the N-terminal, extracellular domain region of human Secretory phospholipase A2 receptor (PLA2R1).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse
PLA2R1, CLEC13C
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit