Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLAC9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579825
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human placenta tissue.
By microarray assay to search for mouse genes enriched in placenta, Plac9 was identified as a placenta-enriched gene expressed in mouse 16-cell morula cDNA. By database analysis, human PLAC9, which encodes a deduced 97-amino acid protein, was identified. The deduced 108-amino acid mouse protein has a predicted signal peptide, a coiled-coil domain, and shares 73% identity with human PLAC9. Northern blot analysis of mouse placenta and embryo RNA at 12.5 and 18.5 days postcoitum (dpc) detected predominant expression in placenta.
Specifications
PLAC9 | |
Polyclonal | |
Unconjugated | |
PLAC9 | |
hypothetical protein LOC510496; PLAC9; Plac9a; placenta associated 9; placenta specific 9; placenta specific 9a; placenta-specific 9; placenta-specific protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219348 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
500 μg/mL | |
PBS with 4 mg trehalose and 0.05 mg sodium azide | |
Q5JTB6 | |
PLAC9 | |
E. coli-derived human PLAC9 recombinant protein (Position: R30-L81). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction