Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.

$142.00 - $492.50

Specifications

Antigen Placental Alk. Phosphatase (PLAP) Ab-4
Clone 8A9
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MS734R7
View Documents
Thermo Scientific™
MS734R7
7mL Each for $321.50
Add to cart
 
MS734S
View Documents
Thermo Scientific™
MS734S
1mL Each for $492.50
Add to cart
 
MS734S0
View Documents
Thermo Scientific™
MS734S0
100μL Each for $142.00
Add to cart
 
MS734S1
View Documents
Thermo Scientific™
MS734S1
500μL Each for $354.50
Add to cart
 
MS734R1
View Documents
Thermo Scientific™
MS734R1
1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states with the Thermo Scientific™ Placental Alk. Phosphatase (PLAP) Ab-4, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: 8A9

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified human Placental Alkaline Phosphatase (PLAP)

Molecular Weight: 60-70kDa

Positive Control: Placenta or seminoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Placental Alk. Phosphatase (PLAP) Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Primary
IgG2a kappa
8A9
Monoclonal
Murine
Cancer and Tumor Biology
Purified human Placental Alkaline Phosphatase (PLAP)
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit