Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Alkaline Phosphatase Monoclonal Antibody (5B1), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MIA1802

Catalog No. PIMIA1802


Add to Cart

Description

Description

MIA1802 targets Alkaline Phosphatase in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIA1802 immunogen is purified human placenta ALP. MIA1802 detects Alkaline Phosphatase which has a predicted molecular weight of approximately 59 kDa. MIA1802 was formerly sold as a Seradyn product.

Placental Alkaline Phosphatase Monoclonal antibody specifically detects Placental Alkaline Phosphatase in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmune Assays (RIA).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Placental Alkaline Phosphatase
Monoclonal
5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
P05187
ALPP
Purified human placenta ALP
1 mg
Primary
Human
Antibody
IgG1
ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
5B1
Unconjugated
ALPP
Alkaline phosphatase placental type; Alkaline phosphatase Regan isozyme; alkaline phosphatase, placental; alkaline phosphatase, placental type; alkaline phosphomonoesterase; ALP; Alp1; ALPP; AP-TNAP; DOA1; FLJ11281; FLJ40094; FLJ61142; FLJ93059; Germ-cell alkaline phosphatase; glycerophosphatase; HOPS; MGC161443; MGC167935; nagao Isozyme; P PLAP; PALP; PLA2P; PLAA; placental alkaline phosphatase 1; placental heat-stable alkaline phosphatase; PLAP; PLAP1; PLAP-1; rCG23846-like; Regan isozyme; TNAP; TNSALP
Mouse
Protein A
RUO
250
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit