Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.

Manufacturer:  Thermo Scientific™ RB9067P1

 View more versions of this product

Catalog No. RB9067P1


Add to cart

Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.

Human placental lactogen (hPL) is known to originate in the syncytiotrophoblasts. Ultrastructural localization indicates that placental lactogen is stored in small morphologically distinct cytoplasmic granules within these cells and has a different secretory mechanism from other hormonal peptides such as human chorionic gonadotropin. The level of expression of placental lactogen through pregnancy increases gradually to term.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Purified human placental lactogen

Molecular Weight: 21kDa

Positive Control: hPL recombinant protein, Placenta

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
Specifications

Specifications

Placental Lactogen (hPL)
Polyclonal
200µg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
IVD
Primary
Polyclonal
Immunohistochemistry (Paraffin), Western Blot
Polyclonal
Unconjugated
Concentrated
Purified human placental lactogen
500μL
Steroids Hormones and Growth Factors
Primary
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit