Learn More
Aviva Systems Biology Corp PLAGL1 Antibody - N-terminal region (AVARP00029_P050) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP00029_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLAGL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLAGL1. Alternate names: DKFZp781P1017, LOT1, MGC126275, MGC126276, ZAC, ZAC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62457