Learn More
PLAGL1 Mouse anti-Human, Alexa Fluor 647, Clone: 8D8C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237343AF647
Catalog No. NBP237343Y
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PLAGL1
8D8C5
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
PLAGL1
Purified recombinant fragment of human PLAGL1 (AA: 118-222) expressed in E. Coli.
Protein A or G purified
Apoptosis, Neuronal Stem Cell Markers, Stem Cell Markers
Primary
5325
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp781P1017, LOT-1, LOT1ZACLost on transformation 1, MGC126275, MGC126276, PLAG-like 1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, ZAC tumor supressor, ZAC1, zinc finger protein PLAGL1
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human