Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plakophilin 1 Antibody (10B2), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254526APCCY7
Description
Plakophilin 1 Monoclonal specifically detects Plakophilin 1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Plakophilin 1 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
PKP1 | |
N-terminus of human Plakophilin 1 protein (Uniprot: Q13835) | |
0.1 mL | |
Primary | |
Recognizes a protein of 75kDa, identified as Plakophilin-1. Its epitope maps between aa 1-27 human Plakophilin-1. Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
10B2 | |
Flow Cytometry | |
B6PBand 6 protein, MGC138829, plakophilin 1 (ectodermal dysplasia/skin fragility syndrome), plakophilin-1 | |
Mouse | |
Protein A or G purified | |
Cellular Markers | |
5317.0 | |
Human, Mouse | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction